The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003955.4 | NP_003946.3 | 2 - 2 | 523 | 101 |
GenBank mRNA | AB004904.1 | - | 1 - 1 | 211 | 101 |
AB006967.1 | - | 1 - 1 | 105 | 101 | |
AF159854.1 | - | 1 - 1 | 105 | 101 | |
AK314212.1 | - | 2 - 2 | 521 | 101 | |
BC060858.1 | - | 2 - 2 | 521 | 101 | |
BT019544.1 | - | 1 - 1 | 105 | 101 | |
CR536497.1 | - | 1 - 1 | 105 | 101 | |
CR541694.1 | - | 1 - 1 | 105 | 101 | |
DA671124.1 | - | 2 - 2 | 523 | 101 |