The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001326475.1 | NP_001313404.1 | 2 - 2 | 561 | 82 |
NM_001326477.1 | NP_001313406.1 | 2 - 2 | 621 | 82 | |
NM_004054.3 | NP_004045.1 | 2 - 2 | 564 | 82 | |
GenBank mRNA | AK223388.1 | - | 2 - 2 | 474 | 82 |
AK290242.1 | - | 2 - 2 | 452 | 82 | |
AK312458.1 | - | 2 - 2 | 452 | 82 | |
BC020742.1 | - | 2 - 2 | 452 | 82 | |
DB013722.1 | - | 2 - 2 | 487 | 82 | |
U28488.1 | - | 2 - 2 | 441 | 82 | |
U62027.1 | - | 2 - 2 | 512 | 82 | |
Z73157.1 | - | 1 - 1 | 360 | 82 |