The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004333.4 | NP_004324.2 | 1 - 2 | 200 | 77 |
XM_005250045.2 | XP_005250102.1 | 1 - 2 | 147 | 77 | |
XM_017012558.1 | XP_016868047.1 | 1 - 2 | 147 | 77 | |
XM_017012559.1 | XP_016868048.1 | 1 - 2 | 147 | 77 | |
GenBank mRNA | AI203435.1 | - | 1 - 2 | 172 | 77 |
AK314346.1 | - | 1 - 2 | 147 | 77 | |
BC101757.1 | - | 1 - 2 | 161 | 77 | |
BC112079.1 | - | 1 - 2 | 161 | 77 | |
DB454480.1 | - | 1 - 2 | 365 | 77 | |
DN996440.1 | - | 1 - 2 | 398 | 77 | |
M95712.2 | - | 1 - 2 | 200 | 77 |