Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ETFDH
Entrez Gene ID
2110
Gene Name
electron transfer flavoprotein dehydrogenase
Gene Aliases
ETFQO, MADD
Chromosome Location
Chr.4: 158672101 - 158708713 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281737.1 NP_001268666.1 5 - 6 904 109
NM_001281738.1 NP_001268667.1 4 - 5 601 109
NM_004453.3 NP_004444.2 6 - 7 1045 109
GenBank mRNA AK293737.1 - 4 - 5 586 109
AK304838.1 - 5 - 6 666 109
AK314803.1 - 6 - 7 1022 109
BC011890.1 - 6 - 7 847 109
BX538129.1 - 7 - 8 717 109
EU093082.1 - 6 - 7 741 109
S69232.1 - 6 - 7 809 109

View Full Product Details