The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004472.2 | NP_004463.1 | 1 - 1 | 1616 | 59 |
GenBank mRNA | U59832.1 | - | 1 - 1 | 1616 | 59 |