The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004496.3 | NP_004487.2 | 1 - 2 | 219 | 74 |
GenBank mRNA | AK313785.1 | - | 1 - 2 | 210 | 74 |
BC033890.1 | - | 1 - 2 | 133 | 74 | |
DA705947.1 | - | 1 - 2 | 221 | 74 | |
U39840.1 | - | 1 - 2 | 158 | 74 |