Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AKAP5
Entrez Gene ID
9495
Gene Name
A-kinase anchoring protein 5
Gene Aliases
AKAP75, AKAP79, H21
Chromosome Location
Chr.14: 64465499 - 64474503 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004857.3 NP_004848.3 2 - 2 717 97
GenBank mRNA AK315336.1 - 2 - 2 718 97
BC057229.1 - 2 - 2 631 97
BC131516.1 - 1 - 1 949 97
M90359.1 - 1 - 1 1636 97

View Full Product Details