The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004907.2 | NP_004898.2 | 2 - 2 | 684 | 105 |
GenBank mRNA | BC003625.2 | - | 1 - 1 | 464 | 105 |
BC072432.1 | - | 2 - 2 | 684 | 105 | |
BC110647.1 | - | 1 - 1 | 1568 | 105 | |
BT019466.1 | - | 1 - 1 | 356 | 105 | |
CR450352.1 | - | 1 - 1 | 356 | 105 | |
M62831.1 | - | 1 - 1 | 456 | 105 |