The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004980.4 | NP_004971.2 | 6 - 7 | 1995 | 70 |
XM_005270851.4 | XP_005270908.1 | 6 - 7 | 1660 | 70 | |
XM_006710629.3 | XP_006710692.1 | 6 - 7 | 1970 | 70 | |
XM_017001244.1 | XP_016856733.1 | 6 - 7 | 1985 | 70 | |
GenBank mRNA | AF048713.1 | - | 5 - 6 | 1515 | 70 |
AF120491.1 | - | 6 - 7 | 2003 | 70 | |
AF187963.1 | - | 5 - 6 | 1587 | 70 | |
AF205857.1 | - | 5 - 6 | 1515 | 70 |