The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243877.1 | NP_001230806.1 | 3 - 3 | 992 | 68 |
NM_001243885.1 | NP_001230814.1 | 2 - 2 | 803 | 68 | |
NM_005749.3 | NP_005740.1 | 2 - 2 | 1006 | 68 | |
GenBank mRNA | AK313904.1 | - | 3 - 3 | 993 | 68 |
AY524047.1 | - | 1 - 1 | 573 | 68 | |
BC031406.1 | - | 1 - 1 | 570 | 68 | |
BC070493.1 | - | 2 - 2 | 994 | 68 | |
BC098415.1 | - | 2 - 2 | 998 | 68 | |
BT019380.1 | - | 1 - 1 | 563 | 68 | |
BT019381.1 | - | 1 - 1 | 563 | 68 | |
CR536487.1 | - | 1 - 1 | 563 | 68 | |
CR541686.1 | - | 1 - 1 | 563 | 68 | |
D38305.1 | - | 1 - 1 | 606 | 68 |