The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005913.2 | NP_005904.1 | 1 - 1 | 711 | 102 |
GenBank mRNA | BC069153.1 | - | 1 - 1 | 490 | 102 |
BC069545.1 | - | 1 - 1 | 489 | 102 | |
BC095531.1 | - | 1 - 1 | 714 | 102 |