The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006278.2 | NP_006269.1 | 1 - 2 | 411 | 91 |
GenBank mRNA | X74570.1 | - | 1 - 2 | 104 | 91 |