Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCT4
Entrez Gene ID
10575
Gene Name
chaperonin containing TCP1 subunit 4
Gene Aliases
CCT-DELTA, Cctd, SRB
Chromosome Location
Chr.2: 61868127 - 61888671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256721.1 NP_001243650.1 11 - 12 1562 79
NM_006430.3 NP_006421.2 12 - 13 1652 79
GenBank mRNA AA136129.1 - 2 - 3 79
AF026291.1 - 12 - 13 1590 79
AK129860.1 - 12 - 13 1561 79
AK290538.1 - 12 - 13 1619 79
AK294665.1 - 11 - 12 1396 79
AK295330.1 - 12 - 13 1425 79
AK303082.1 - 11 - 12 1547 79
AK307856.1 - 13 - 14 1587 80
AK308942.1 - 12 - 13 1569 79
AK312586.1 - 12 - 13 1594 79
AK315975.1 - 13 - 14 1828 79
AL555753.3 - 2 - 3 79
BC014676.1 - 5 - 6 622 79
BC106933.1 - 12 - 13 1585 79
BC106934.1 - 12 - 13 1588 79
U38846.1 - 12 - 13 1488 79

View Full Product Details