Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39870120-39870633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAAM2
Entrez Gene ID
23500
Gene Name
dishevelled associated activator of morphogenesis 2
Gene Aliases
dJ90A20A.1
UniGene
Hs.357128
Gene Chromosome Location
Chr.6: 39792366-39904877 on Build GRCh38
Transcripts
NM_001201427.1 NM_015345.3 XM_006715039.3 XM_006715040.3 XM_006715042.2 XM_006715043.2 XM_006715045.3 XM_006715046.3 XM_017010630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6885717 c.930G>A p.Q310Q Chr.6: 39870396-39870396 on Build GRCh38
6402464 c.971T>A p.L324Q Chr.6: 39870437-39870437 on Build GRCh38
5710116 c.889C>T p.R297C Chr.6: 39870355-39870355 on Build GRCh38
5794563 c.890G>A p.R297H Chr.6: 39870356-39870356 on Build GRCh38
4775823 c.977+8C>T p.? Chr.6: 39870451-39870451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574793504 Chr.6: 39870512-39870512 on Build GRCh38
rs754862494 Chr.6: 39870293-39870293 on Build GRCh38
rs887090078 Chr.6: 39870587-39870587 on Build GRCh38
rs532710553 Chr.6: 39870338-39870338 on Build GRCh38
rs78880432 Chr.6: 39870585-39870585 on Build GRCh38
rs776399197 Chr.6: 39870414-39870414 on Build GRCh38
rs557124617 Chr.6: 39870583-39870583 on Build GRCh38
rs772990860 Chr.6: 39870571-39870571 on Build GRCh38
rs762364231 Chr.6: 39870437-39870437 on Build GRCh38
rs964177492 Chr.6: 39870474-39870474 on Build GRCh38
rs964230451 Chr.6: 39870603-39870603 on Build GRCh38
rs549473617 Chr.6: 39870216-39870216 on Build GRCh38
rs552429229 Chr.6: 39870609-39870609 on Build GRCh38
rs1019189651 Chr.6: 39870519-39870519 on Build GRCh38
rs374061585 Chr.6: 39870383-39870383 on Build GRCh38
rs926562956 Chr.6: 39870582-39870583 on Build GRCh38
rs898550504 Chr.6: 39870356-39870356 on Build GRCh38
rs995610152 Chr.6: 39870367-39870367 on Build GRCh38
rs975113590 Chr.6: 39870228-39870228 on Build GRCh38
rs768190596 Chr.6: 39870458-39870458 on Build GRCh38
rs965450653 Chr.6: 39870537-39870537 on Build GRCh38
rs370816394 Chr.6: 39870309-39870309 on Build GRCh38
rs1002078681 Chr.6: 39870305-39870305 on Build GRCh38
rs975082235 Chr.6: 39870541-39870541 on Build GRCh38
rs987710761 Chr.6: 39870145-39870145 on Build GRCh38
rs537461717 Chr.6: 39870564-39870564 on Build GRCh38
rs376342216 Chr.6: 39870268-39870268 on Build GRCh38
rs367983386 Chr.6: 39870459-39870459 on Build GRCh38
rs747434941 Chr.6: 39870452-39870452 on Build GRCh38
rs35016293 Chr.6: 39870365-39870366 on Build GRCh38
rs1027969677 Chr.6: 39870560-39870560 on Build GRCh38
rs949301943 Chr.6: 39870279-39870279 on Build GRCh38
rs759121825 Chr.6: 39870415-39870415 on Build GRCh38
rs569621035 Chr.6: 39870240-39870240 on Build GRCh38
rs1019815403 Chr.6: 39870597-39870597 on Build GRCh38
rs184450403 Chr.6: 39870461-39870461 on Build GRCh38
rs905012091 Chr.6: 39870302-39870302 on Build GRCh38
rs925462006 Chr.6: 39870515-39870515 on Build GRCh38
rs777079374 Chr.6: 39870497-39870497 on Build GRCh38
rs913527301 Chr.6: 39870561-39870561 on Build GRCh38
rs746561768 Chr.6: 39870390-39870390 on Build GRCh38
rs576804226 Chr.6: 39870584-39870584 on Build GRCh38
rs1052496792 Chr.6: 39870277-39870277 on Build GRCh38
rs965056235 Chr.6: 39870215-39870215 on Build GRCh38
rs566309443 Chr.6: 39870325-39870325 on Build GRCh38
rs552880915 Chr.6: 39870267-39870267 on Build GRCh38
rs772017388 Chr.6: 39870355-39870355 on Build GRCh38
rs555066503 Chr.6: 39870506-39870506 on Build GRCh38
rs539385292 Chr.6: 39870261-39870261 on Build GRCh38
rs925072462 Chr.6: 39870259-39870259 on Build GRCh38
rs563329976 Chr.6: 39870195-39870195 on Build GRCh38
rs370812440 Chr.6: 39870431-39870431 on Build GRCh38
rs768996183 Chr.6: 39870416-39870416 on Build GRCh38
rs774613617 Chr.6: 39870435-39870435 on Build GRCh38
rs921005645 Chr.6: 39870260-39870260 on Build GRCh38
rs912702147 Chr.6: 39870175-39870175 on Build GRCh38
rs746546828 Chr.6: 39870594-39870594 on Build GRCh38
rs770562217 Chr.6: 39870391-39870391 on Build GRCh38
rs1026451158 Chr.6: 39870499-39870499 on Build GRCh38
rs1009586946 Chr.6: 39870513-39870513 on Build GRCh38
rs370351113 Chr.6: 39870460-39870460 on Build GRCh38
rs913707674 Chr.6: 39870278-39870278 on Build GRCh38
rs373590256 Chr.6: 39870593-39870593 on Build GRCh38

View Full Product Details