The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014060.2 | NP_054779.1 | 1 - 2 | 350 | 83 |
GenBank mRNA | AB034206.1 | - | 1 - 2 | 259 | 83 |
AK311993.1 | - | 1 - 2 | 312 | 83 | |
BC001013.2 | - | 1 - 2 | 73 | 83 | |
BC095461.1 | - | - | 125 | 83 | |
BP226595.1 | - | 1 - 2 | 350 | 83 |