The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308155.1 | NP_001295084.1 | 13 - 14 | 2414 | 99 |
NM_014498.4 | NP_055313.1 | 14 - 15 | 2498 | 99 | |
XM_005247364.2 | XP_005247421.1 | 14 - 15 | 2500 | 99 | |
XM_005247365.2 | XP_005247422.1 | 13 - 14 | 2416 | 99 | |
XM_017006183.1 | XP_016861672.1 | 13 - 14 | 1896 | 99 | |
GenBank mRNA | AK307557.1 | - | 14 - 15 | 2203 | 99 |
U55853.1 | - | 14 - 15 | 1871 | 99 |