The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014908.3 | NP_055723.1 | 1 - 1 | 1424 | 103 |
GenBank mRNA | AB029017.1 | - | 1 - 1 | 1310 | 103 |
AY358759.1 | - | 1 - 1 | 1279 | 103 | |
BC035556.1 | - | 1 - 1 | 1277 | 103 | |
KU178553.1 | - | 1 - 1 | 994 | 103 |