The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303533.1 | NP_001290462.1 | 8 - 9 | 929 | 64 |
NM_001303534.1 | NP_001290463.1 | 8 - 9 | 1024 | 64 | |
NM_016339.4 | NP_057423.1 | 8 - 9 | 1209 | 64 | |
XM_006721932.3 | XP_006721995.1 | 8 - 9 | 1703 | 64 | |
XM_006721935.3 | XP_006721998.1 | 9 - 10 | 980 | 64 | |
XM_006721937.3 | XP_006722000.1 | 8 - 9 | 2805 | 64 | |
XM_006721938.3 | XP_006722001.1 | 5 - 6 | 596 | 64 | |
GenBank mRNA | AF117946.1 | - | 8 - 9 | 844 | 64 |
AK294642.1 | - | 4 - 5 | 831 | 64 | |
AK304160.1 | - | 8 - 9 | 1024 | 64 | |
AK313492.1 | - | 8 - 9 | 884 | 64 | |
BC029956.1 | - | 8 - 9 | 1703 | 64 | |
BC115374.1 | - | 6 - 7 | 738 | 64 | |
BC115375.1 | - | 6 - 7 | 738 | 64 |