Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43597698-43598244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLH
Entrez Gene ID
5429
Gene Name
DNA polymerase eta
Gene Aliases
RAD30, RAD30A, XP-V, XPV
UniGene
Hs.655467
Gene Chromosome Location
Chr.6: 43576141-43620523 on Build GRCh38
Transcripts
NM_001291969.1 NM_001291970.1 NM_006502.2 XM_005249186.3 XM_011514698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1444754 c.583G>A p.V195M Chr.6: 43597788-43597788 on Build GRCh38
5549105 c.641C>T p.A214V Chr.6: 43597846-43597846 on Build GRCh38
7094850 c.525C>T p.L175L Chr.6: 43597730-43597730 on Build GRCh38
1131852 c.526G>C p.D176H Chr.6: 43597731-43597731 on Build GRCh38
7085645 c.500_540del41 p.K168fs*1 Chr.6: 43597705-43597745 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961344756 Chr.6: 43598065-43598066 on Build GRCh38
rs561673278 Chr.6: 43598103-43598103 on Build GRCh38
rs760104150 Chr.6: 43597776-43597776 on Build GRCh38
rs748225581 Chr.6: 43597724-43597724 on Build GRCh38
rs887492114 Chr.6: 43597995-43597995 on Build GRCh38
rs1049938684 Chr.6: 43597887-43597887 on Build GRCh38
rs755012512 Chr.6: 43598003-43598003 on Build GRCh38
rs866807275 Chr.6: 43597967-43597967 on Build GRCh38
rs2307456 Chr.6: 43597831-43597831 on Build GRCh38
rs1044736898 Chr.6: 43597994-43597994 on Build GRCh38
rs756575357 Chr.6: 43597761-43597761 on Build GRCh38
rs947749458 Chr.6: 43597993-43597993 on Build GRCh38
rs751080449 Chr.6: 43597856-43597856 on Build GRCh38
rs573515726 Chr.6: 43598134-43598134 on Build GRCh38
rs751252858 Chr.6: 43598108-43598108 on Build GRCh38
rs971414692 Chr.6: 43598030-43598030 on Build GRCh38
rs779127982 Chr.6: 43597783-43597783 on Build GRCh38
rs770881756 Chr.6: 43597815-43597815 on Build GRCh38
rs757995181 Chr.6: 43597979-43597979 on Build GRCh38
rs772090914 Chr.6: 43597800-43597800 on Build GRCh38
rs575829098 Chr.6: 43597960-43597960 on Build GRCh38
rs56056074 Chr.6: 43597875-43597875 on Build GRCh38
rs775225254 Chr.6: 43597839-43597839 on Build GRCh38
rs370763770 Chr.6: 43598065-43598065 on Build GRCh38
rs754158551 Chr.6: 43597870-43597870 on Build GRCh38
rs971195455 Chr.6: 43597751-43597751 on Build GRCh38
rs748244166 Chr.6: 43597790-43597790 on Build GRCh38
rs758591272 Chr.6: 43597901-43597901 on Build GRCh38
rs150901869 Chr.6: 43597730-43597730 on Build GRCh38
rs1031040918 Chr.6: 43598101-43598101 on Build GRCh38
rs985110671 Chr.6: 43598147-43598147 on Build GRCh38
rs1015022354 Chr.6: 43598024-43598024 on Build GRCh38
rs776345153 Chr.6: 43597817-43597817 on Build GRCh38
rs774355110 Chr.6: 43597719-43597719 on Build GRCh38
rs986503269 Chr.6: 43598153-43598153 on Build GRCh38
rs747806616 Chr.6: 43597738-43597739 on Build GRCh38
rs981258983 Chr.6: 43598051-43598051 on Build GRCh38
rs577721367 Chr.6: 43597928-43597928 on Build GRCh38
rs2307457 Chr.6: 43597959-43597959 on Build GRCh38
rs1005939285 Chr.6: 43598015-43598015 on Build GRCh38
rs935218807 Chr.6: 43597976-43597976 on Build GRCh38
rs759369209 Chr.6: 43597821-43597821 on Build GRCh38
rs144656482 Chr.6: 43597948-43597948 on Build GRCh38
rs903338844 Chr.6: 43598202-43598204 on Build GRCh38
rs536476976 Chr.6: 43598160-43598161 on Build GRCh38
rs534738892 Chr.6: 43597872-43597872 on Build GRCh38
rs200691612 Chr.6: 43597857-43597857 on Build GRCh38
rs1023012195 Chr.6: 43598023-43598023 on Build GRCh38
rs752880762 Chr.6: 43597898-43597898 on Build GRCh38
rs773080170 Chr.6: 43597731-43597731 on Build GRCh38
rs368152403 Chr.6: 43597767-43597767 on Build GRCh38
rs768147472 Chr.6: 43597749-43597749 on Build GRCh38
rs997764252 Chr.6: 43598069-43598069 on Build GRCh38
rs912319785 Chr.6: 43598149-43598149 on Build GRCh38
rs776458288 Chr.6: 43597732-43597732 on Build GRCh38
rs1015349598 Chr.6: 43598053-43598053 on Build GRCh38
rs947461177 Chr.6: 43597951-43597951 on Build GRCh38
rs763654639 Chr.6: 43597843-43597843 on Build GRCh38
rs764927842 Chr.6: 43597733-43597733 on Build GRCh38
rs541030892 Chr.6: 43598146-43598146 on Build GRCh38
rs943748315 Chr.6: 43598150-43598150 on Build GRCh38
rs755224547 Chr.6: 43597779-43597779 on Build GRCh38
rs369844648 Chr.6: 43597778-43597778 on Build GRCh38
rs560020212 Chr.6: 43598202-43598203 on Build GRCh38
rs762484795 Chr.6: 43597747-43597747 on Build GRCh38
rs766918319 Chr.6: 43597722-43597722 on Build GRCh38
rs752215374 Chr.6: 43597744-43597744 on Build GRCh38
rs766907046 Chr.6: 43597861-43597861 on Build GRCh38
rs896144065 Chr.6: 43597981-43597981 on Build GRCh38
rs777719634 Chr.6: 43597809-43597809 on Build GRCh38
rs569896052 Chr.6: 43597737-43597737 on Build GRCh38
rs762570589 Chr.6: 43597842-43597842 on Build GRCh38
rs777380961 Chr.6: 43598048-43598048 on Build GRCh38
rs750908181 Chr.6: 43597757-43597757 on Build GRCh38
rs903457801 Chr.6: 43597975-43597975 on Build GRCh38

View Full Product Details