Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FUT4
Entrez Gene ID
2526
Gene Name
fucosyltransferase 4
Gene Aliases
CD15, ELFT, FCT3A, FUC-TIV, FUTIV, LeX, SSEA-1
Chromosome Location
Chr.11: 94543851 - 94549898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002033.3 NP_002024.1 1 - 1 1893 89
GenBank mRNA BC136373.1 - 1 - 1 1773 89
BC136374.1 - 1 - 1 1773 89
M58596.1 - 1 - 1 1292 89
M58597.1 - 1 - 1 1783 89

Gene Symbol
LOC105369438
Entrez Gene ID
105369438
Gene Name
uncharacterized LOC105369438
Gene Aliases
-
Chromosome Location
Chr.11: 94545332 - 94740355 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_135093.1 5 - 5 89
GenBank mRNA BI522712.1 - 5 - 5 89

Gene Symbol
PIWIL4
Entrez Gene ID
143689
Gene Name
piwi like RNA-mediated gene silencing 4
Gene Aliases
HIWI2, MIWI2
Chromosome Location
Chr.11: 94567308 - 94621421 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CR749642.1 - 1 - 1 151 89

View Full Product Details