The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014675.4 | NP_055490.4 | 24 - 25 | 3772 | 100 |
XM_006711058.3 | XP_006711121.1 | 23 - 24 | 3563 | 100 | |
XM_011542468.1 | XP_011540770.1 | 24 - 25 | 3749 | 100 | |
XM_011542470.2 | XP_011540772.1 | 24 - 25 | 3749 | 100 | |
XM_017002913.1 | XP_016858402.1 | 24 - 25 | 3749 | 100 | |
XM_017002914.1 | XP_016858403.1 | 24 - 25 | 3749 | 100 | |
XM_017002915.1 | XP_016858404.1 | 21 - 22 | 3257 | 100 | |
GenBank mRNA | AB007914.2 | - | 22 - 23 | 3411 | 100 |
BC126911.1 | - | 21 - 22 | 3293 | 100 | |
BC126912.1 | - | 21 - 22 | 3293 | 100 | |
DQ139275.1 | - | 11 - 12 | 3129 | 100 |