The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032316.3 | NP_115692.1 | 1 - 2 | 215 | 99 |
GenBank mRNA | AJ299740.1 | - | 1 - 2 | 187 | 99 |
AK094248.1 | - | 1 - 2 | 155 | 99 | |
AK301846.1 | - | 1 - 2 | 218 | 99 | |
AK313073.1 | - | 1 - 2 | 212 | 99 | |
BC005050.1 | - | 1 - 2 | 132 | 99 | |
BC050653.1 | - | 1 - 2 | 179 | 99 |