The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001280547.1 | NP_001267476.1 | 4 - 5 | 929 | 84 |
NM_001280548.1 | NP_001267477.1 | 4 - 5 | 929 | 84 | |
NM_001280549.1 | NP_001267478.1 | 4 - 5 | 929 | 84 | |
NM_001280550.1 | NP_001267479.1 | 4 - 5 | 929 | 84 | |
NM_001280551.1 | NP_001267480.1 | 3 - 4 | 763 | 84 | |
NM_001280552.1 | NP_001267481.1 | 4 - 5 | 929 | 84 | |
NM_001280555.1 | NP_001267484.1 | 3 - 4 | 731 | 84 | |
NM_001280556.1 | NP_001267485.1 | 3 - 4 | 763 | 84 | |
NM_016734.2 | NP_057953.1 | 4 - 5 | 929 | 84 | |
NR_103999.1 | 4 - 5 | 929 | 84 | ||
NR_104000.1 | 4 - 5 | 929 | 84 | ||
XM_005251481.4 | XP_005251538.1 | 4 - 5 | 1550 | 84 | |
XM_011517896.2 | XP_011516198.1 | 4 - 5 | 2399 | 84 | |
XM_011517897.2 | XP_011516199.1 | 4 - 5 | 1554 | 84 | |
GenBank mRNA | AF080573.1 | - | 3 - 4 | 299 | 84 |
AY463952.1 | - | 4 - 5 | 491 | 84 | |
AY463953.1 | - | 4 - 5 | 491 | 84 | |
AY463954.1 | - | 4 - 5 | 491 | 84 | |
AY463955.1 | - | 4 - 5 | 491 | 84 | |
AY463956.1 | - | 4 - 5 | 491 | 84 | |
AY463957.1 | - | 4 - 5 | 491 | 84 | |
FJ626415.1 | - | 3 - 4 | 312 | 84 | |
FJ626418.1 | - | 2 - 3 | 114 | 84 | |
FJ626419.1 | - | 3 - 4 | 84 | ||
FJ626420.1 | - | 3 - 4 | 315 | 84 | |
FJ626421.1 | - | 4 - 5 | 481 | 84 | |
FJ626424.1 | - | 3 - 4 | 283 | 84 | |
FJ626425.1 | - | 4 - 5 | 481 | 84 | |
M96944.1 | - | 4 - 5 | 556 | 84 |