Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE4D
Entrez Gene ID
5144
Gene Name
phosphodiesterase 4D
Gene Aliases
ACRDYS2, DPDE3, HSPDE4D, PDE43, PDE4DN2, STRK1
Chromosome Location
Chr.5: 58969038 - 60488098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104631.1 NP_001098101.1 3 - 4 77
NM_001165899.1 NP_001159371.1 5 - 6 77
NM_001197218.1 NP_001184147.1 3 - 4 77
NM_001197219.1 NP_001184148.1 3 - 4 77
NM_001197220.1 NP_001184149.1 3 - 4 77
NM_006203.4 NP_006194.2 3 - 4 77
XM_005248537.3 XP_005248594.1 3 - 4 77
XM_005248538.4 XP_005248595.1 4 - 5 77
XM_011543469.1 XP_011541771.1 6 - 7 77
XM_011543470.2 XP_011541772.1 6 - 7 77
XM_011543471.1 XP_011541773.1 5 - 6 77
XM_011543473.1 XP_011541775.1 5 - 6 77
XM_011543477.2 XP_011541779.1 5 - 6 77
XM_017009565.1 XP_016865054.1 6 - 7 77
XM_017009566.1 XP_016865055.1 6 - 7 77
XM_017009567.1 XP_016865056.1 4 - 5 77
XM_017009568.1 XP_016865057.1 6 - 7 77
XM_017009569.1 XP_016865058.1 5 - 6 77
GenBank mRNA AF012073.1 - 3 - 4 77
AF536976.1 - 4 - 5 77
AF536977.1 - 3 - 4 77
AJ250854.1 - 3 - 4 77
AJ250855.1 - 3 - 4 77
AY245866.1 - 5 - 6 77
AY245867.1 - 3 - 4 77
AY388960.1 - 3 - 4 77
BC008390.1 - 3 - 4 77
BT007398.1 - 3 - 4 77
L20970.1 - 3 - 4 77
U02882.1 - 2 - 3 77
U50159.1 - 3 - 4 350 77

View Full Product Details