Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VEPH1
Entrez Gene ID
79674
Gene Name
ventricular zone expressed PH domain containing 1
Gene Aliases
MELT, VEPH
Chromosome Location
Chr.3: 157259742 - 157503626 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167912.1 NP_001161384.1 10 - 11 2102 99
NM_024621.2 NP_078897.2 10 - 11 2293 99
XM_011513134.2 XP_011511436.1 9 - 10 2036 99
XM_011513135.1 XP_011511437.1 10 - 11 1932 99
GenBank mRNA AB051479.1 - 10 - 11 2102 99
AK027625.1 - 10 - 11 2191 99
AL713656.1 - 9 - 10 1890 99
BC026235.1 - 2 - 3 415 99
BC042159.1 - 3 - 4 752 99
BC101660.1 - 9 - 10 1929 99
BC113555.1 - 9 - 10 1929 99

View Full Product Details