The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152643.6 | NP_689856.6 | 3 - 4 | 364 | 68 |
XM_017016858.1 | XP_016872347.1 | 3 - 4 | 732 | 68 | |
XM_017016859.1 | XP_016872348.1 | 3 - 4 | 735 | 68 | |
GenBank mRNA | AB000781.1 | - | 2 - 3 | 260 | 68 |
AB051555.1 | - | 2 - 3 | 458 | 68 | |
AK090457.1 | - | 3 - 4 | 327 | 68 |