The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032430.1 | NP_115806.1 | 3 - 4 | 598 | 95 |
GenBank mRNA | AB058714.1 | - | 2 - 3 | 133 | 95 |
AF479826.1 | - | 5 - 6 | 496 | 95 | |
AF479827.1 | - | 3 - 4 | 598 | 95 | |
AL834275.1 | - | 3 - 4 | 221 | 95 | |
AY458602.1 | - | 3 - 4 | 370 | 95 | |
AY505124.1 | - | 3 - 4 | 321 | 95 | |
HQ830199.1 | - | 3 - 4 | 343 | 95 |