The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052911.2 | NP_443143.2 | 8 - 9 | 2889 | 87 |
XM_011525798.1 | XP_011524100.1 | 8 - 9 | 3019 | 87 | |
GenBank mRNA | AB067498.1 | - | 8 - 9 | 2825 | 87 |
AK090579.1 | - | 5 - 6 | 647 | 87 | |
BC089426.1 | - | 5 - 6 | 2309 | 87 |