Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66638174-66638681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD7
Entrez Gene ID
154881
Gene Name
potassium channel tetramerization domain containing 7
Gene Aliases
CLN14, EPM3
UniGene
Hs.546627
Gene Chromosome Location
Chr.7: 66628881-66643229 on Build GRCh38
Transcripts
NM_001167961.2 NM_153033.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6692118 c.368G>A p.R123Q Chr.7: 66638306-66638306 on Build GRCh38
3640113 c.315-1G>A p.? Chr.7: 66638252-66638252 on Build GRCh38
6692119 c.388C>A p.Q130K Chr.7: 66638326-66638326 on Build GRCh38
3367006 c.490A>G p.K164E Chr.7: 66638428-66638428 on Build GRCh38
453302 c.335G>T p.R112L Chr.7: 66638273-66638273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531332669 Chr.7: 66638551-66638551 on Build GRCh38
rs1006793740 Chr.7: 66638549-66638549 on Build GRCh38
rs778046189 Chr.7: 66638218-66638218 on Build GRCh38
rs746772828 Chr.7: 66638244-66638244 on Build GRCh38
rs199624315 Chr.7: 66638300-66638300 on Build GRCh38
rs779291721 Chr.7: 66638203-66638203 on Build GRCh38
rs765235486 Chr.7: 66638396-66638396 on Build GRCh38
rs989198210 Chr.7: 66638540-66638540 on Build GRCh38
rs886648324 Chr.7: 66638518-66638518 on Build GRCh38
rs182290415 Chr.7: 66638517-66638517 on Build GRCh38
rs755121011 Chr.7: 66638468-66638468 on Build GRCh38
rs370881492 Chr.7: 66638243-66638243 on Build GRCh38
rs75672175 Chr.7: 66638229-66638229 on Build GRCh38
rs765083971 Chr.7: 66638318-66638318 on Build GRCh38
rs199604642 Chr.7: 66638299-66638299 on Build GRCh38
rs762715865 Chr.7: 66638506-66638506 on Build GRCh38
rs561779629 Chr.7: 66638608-66638608 on Build GRCh38
rs145152329 Chr.7: 66638280-66638280 on Build GRCh38
rs976891930 Chr.7: 66638230-66638230 on Build GRCh38
rs761707159 Chr.7: 66638395-66638395 on Build GRCh38
rs374072904 Chr.7: 66638349-66638349 on Build GRCh38
rs746365138 Chr.7: 66638206-66638206 on Build GRCh38
rs754958440 Chr.7: 66638340-66638340 on Build GRCh38
rs772468381 Chr.7: 66638226-66638226 on Build GRCh38
rs774026720 Chr.7: 66638273-66638273 on Build GRCh38
rs574847246 Chr.7: 66638377-66638377 on Build GRCh38
rs367608044 Chr.7: 66638305-66638305 on Build GRCh38
rs746540809 Chr.7: 66638382-66638382 on Build GRCh38
rs147604268 Chr.7: 66638651-66638651 on Build GRCh38
rs748562227 Chr.7: 66638268-66638268 on Build GRCh38
rs372103637 Chr.7: 66638336-66638336 on Build GRCh38
rs780330313 Chr.7: 66638213-66638213 on Build GRCh38
rs145238250 Chr.7: 66638322-66638322 on Build GRCh38
rs1003054690 Chr.7: 66638283-66638283 on Build GRCh38
rs751720071 Chr.7: 66638338-66638338 on Build GRCh38
rs936496800 Chr.7: 66638369-66638369 on Build GRCh38
rs191592577 Chr.7: 66638482-66638482 on Build GRCh38
rs527385175 Chr.7: 66638630-66638630 on Build GRCh38
rs767984903 Chr.7: 66638385-66638385 on Build GRCh38
rs764148503 Chr.7: 66638306-66638306 on Build GRCh38
rs761672292 Chr.7: 66638313-66638313 on Build GRCh38
rs879025829 Chr.7: 66638439-66638439 on Build GRCh38
rs74362924 Chr.7: 66638488-66638488 on Build GRCh38
rs546503242 Chr.7: 66638494-66638494 on Build GRCh38
rs555039386 Chr.7: 66638343-66638343 on Build GRCh38
rs751667494 Chr.7: 66638463-66638463 on Build GRCh38
rs397720861 Chr.7: 66638452-66638453 on Build GRCh38
rs769991459 Chr.7: 66638272-66638272 on Build GRCh38
rs781613111 Chr.7: 66638453-66638454 on Build GRCh38
rs747057275 Chr.7: 66638218-66638218 on Build GRCh38
rs938944327 Chr.7: 66638516-66638516 on Build GRCh38
rs541499610 Chr.7: 66638603-66638603 on Build GRCh38
rs554855474 Chr.7: 66638576-66638576 on Build GRCh38
rs900567360 Chr.7: 66638586-66638586 on Build GRCh38
rs978399798 Chr.7: 66638554-66638554 on Build GRCh38
rs140932942 Chr.7: 66638325-66638325 on Build GRCh38
rs758573631 Chr.7: 66638205-66638205 on Build GRCh38
rs753658170 Chr.7: 66638307-66638307 on Build GRCh38
rs747385800 Chr.7: 66638359-66638360 on Build GRCh38
rs886645555 Chr.7: 66638445-66638445 on Build GRCh38
rs756458562 Chr.7: 66638572-66638572 on Build GRCh38
rs775109352 Chr.7: 66638378-66638378 on Build GRCh38
rs768768596 Chr.7: 66638265-66638265 on Build GRCh38
rs915073290 Chr.7: 66638550-66638550 on Build GRCh38
rs978040042 Chr.7: 66638631-66638631 on Build GRCh38
rs758037249 Chr.7: 66638218-66638233 on Build GRCh38
rs57580125 Chr.7: 66638448-66638449 on Build GRCh38
rs387907262 Chr.7: 66638281-66638281 on Build GRCh38
rs932417352 Chr.7: 66638249-66638249 on Build GRCh38
rs781725855 Chr.7: 66638341-66638341 on Build GRCh38
rs796052686 Chr.7: 66638394-66638394 on Build GRCh38
rs749618351 Chr.7: 66638368-66638368 on Build GRCh38
rs555342583 Chr.7: 66638606-66638606 on Build GRCh38
rs906179618 Chr.7: 66638502-66638502 on Build GRCh38
rs754349657 Chr.7: 66638659-66638659 on Build GRCh38
rs187113750 Chr.7: 66638457-66638457 on Build GRCh38
rs540783571 Chr.7: 66638424-66638424 on Build GRCh38
rs186629381 Chr.7: 66638580-66638580 on Build GRCh38
rs778155909 Chr.7: 66638357-66638357 on Build GRCh38
rs776661654 Chr.7: 66638386-66638386 on Build GRCh38
rs368833598 Chr.7: 66638507-66638507 on Build GRCh38
rs773209661 Chr.7: 66638403-66638403 on Build GRCh38
rs374072494 Chr.7: 66638654-66638654 on Build GRCh38
rs927872891 Chr.7: 66638573-66638573 on Build GRCh38
rs387907263 Chr.7: 66638260-66638260 on Build GRCh38

View Full Product Details