Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK054979.1 - 1 453 Chr. 17 - 79268841 - 79271085 [-] on Build GRCh38 76
GenBankmRNA DA060417.1 - 1 453 Chr. 17 - 79270534 - 79271085 [-] on Build GRCh38 76


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream RNA binding protein, fox-1 homolog 3 RBFOX3 FOX-3, FOX3, HRNBP3, NEUN Sense Chr. - 79089345 - 79611101 on Build GRCh38
Downstream microRNA 4739 MIR4739 mir-4739 436.1kb Sense Chr. - 79707176 - 79707249 on Build GRCh38

View Full Product Details