Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK055008.1 - 1 649 Chr. 14 - 61801713 - 61803634 [+] on Build GRCh38 77


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream small nuclear RNA activating complex polypeptide 1 SNAPC1 PTFgamma, SNAP43 -5.3kb Sense Chr. - 61762357 - 61796428 on Build GRCh38
Downstream synaptotagmin 16 SYT16 CHR14SYT, SYT14L, Strep14, syt14r, yt14r 9.1kb Sense Chr. - 61812156 - 62112825 on Build GRCh38

View Full Product Details