Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100403740-100404231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
UniGene
Hs.105191
Gene Chromosome Location
Chr.7: 100400826-100429239 on Build GRCh38
Transcripts
NM_001258008.1 NM_017984.4 XM_005250479.2 XM_005250480.2 XM_006716035.3 XM_006716036.2 XM_006716037.3 XM_006716038.3 XM_006716040.3 XM_017012379.1 XM_017012380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3662782 c.1318+10A>G p.? Chr.7: 100404168-100404168 on Build GRCh38
5062012 c.1346C>A p.P449Q Chr.7: 100403758-100403758 on Build GRCh38
4464225 c.1320C>T p.D440D Chr.7: 100403784-100403784 on Build GRCh38
1453418 c.1309delG p.E437fs*10 Chr.7: 100404187-100404187 on Build GRCh38
3630732 c.1313G>A p.R438K Chr.7: 100404183-100404183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761995553 Chr.7: 100404156-100404156 on Build GRCh38
rs989895845 Chr.7: 100403867-100403867 on Build GRCh38
rs2158862 Chr.7: 100404173-100404173 on Build GRCh38
rs755043591 Chr.7: 100403809-100403809 on Build GRCh38
rs758975326 Chr.7: 100403780-100403780 on Build GRCh38
rs747796375 Chr.7: 100403801-100403801 on Build GRCh38
rs562401270 Chr.7: 100404085-100404085 on Build GRCh38
rs978280207 Chr.7: 100404163-100404163 on Build GRCh38
rs774381056 Chr.7: 100404132-100404132 on Build GRCh38
rs928128457 Chr.7: 100403959-100403959 on Build GRCh38
rs772923348 Chr.7: 100403809-100403809 on Build GRCh38
rs574204185 Chr.7: 100404086-100404086 on Build GRCh38
rs769466428 Chr.7: 100403804-100403804 on Build GRCh38
rs549115411 Chr.7: 100404167-100404167 on Build GRCh38
rs765252928 Chr.7: 100403835-100403836 on Build GRCh38
rs768855784 Chr.7: 100403796-100403796 on Build GRCh38
rs374009133 Chr.7: 100403808-100403808 on Build GRCh38
rs780475059 Chr.7: 100403787-100403787 on Build GRCh38
rs948116593 Chr.7: 100403945-100403945 on Build GRCh38
rs771003171 Chr.7: 100403835-100403835 on Build GRCh38
rs368172772 Chr.7: 100404207-100404207 on Build GRCh38
rs971836079 Chr.7: 100403929-100403929 on Build GRCh38
rs202188414 Chr.7: 100404194-100404194 on Build GRCh38
rs770934294 Chr.7: 100403964-100403964 on Build GRCh38
rs1025628907 Chr.7: 100403868-100403868 on Build GRCh38
rs747389347 Chr.7: 100403789-100403789 on Build GRCh38
rs751972199 Chr.7: 100404189-100404189 on Build GRCh38
rs776702960 Chr.7: 100404063-100404063 on Build GRCh38
rs780930255 Chr.7: 100403800-100403800 on Build GRCh38
rs1045154933 Chr.7: 100403951-100403951 on Build GRCh38
rs1038460431 Chr.7: 100404090-100404090 on Build GRCh38
rs766600387 Chr.7: 100403767-100403777 on Build GRCh38
rs775329098 Chr.7: 100404154-100404154 on Build GRCh38
rs750510774 Chr.7: 100404160-100404160 on Build GRCh38
rs781458267 Chr.7: 100404206-100404206 on Build GRCh38
rs994681485 Chr.7: 100403853-100403853 on Build GRCh38
rs754047881 Chr.7: 100403780-100403781 on Build GRCh38
rs768175247 Chr.7: 100404156-100404162 on Build GRCh38
rs755480981 Chr.7: 100404205-100404205 on Build GRCh38
rs1047262859 Chr.7: 100404068-100404068 on Build GRCh38
rs939559799 Chr.7: 100404048-100404048 on Build GRCh38
rs887363326 Chr.7: 100404079-100404081 on Build GRCh38
rs759403793 Chr.7: 100404149-100404149 on Build GRCh38
rs200478128 Chr.7: 100404152-100404152 on Build GRCh38
rs748739888 Chr.7: 100403827-100403827 on Build GRCh38
rs1035316083 Chr.7: 100404045-100404045 on Build GRCh38
rs990631217 Chr.7: 100404087-100404087 on Build GRCh38
rs533272289 Chr.7: 100404165-100404165 on Build GRCh38
rs1003457931 Chr.7: 100403972-100403972 on Build GRCh38
rs868341355 Chr.7: 100404030-100404030 on Build GRCh38
rs915103987 Chr.7: 100404111-100404111 on Build GRCh38
rs544603126 Chr.7: 100404091-100404091 on Build GRCh38

View Full Product Details