The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025179.3 | NP_079455.3 | 32 - 32 | 8677 | 91 |
XM_005273164.2 | XP_005273221.1 | 33 - 33 | 8936 | 91 | |
GenBank mRNA | AK055196.1 | - | 1 - 1 | 55 | 91 |
AK125154.1 | - | 1 - 1 | 604 | 91 |