The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008224.1 | NP_001008225.1 | 5 - 6 | 750 | 70 |
NM_018003.2 | NP_060473.2 | 5 - 6 | 527 | 70 | |
XM_005254529.4 | XP_005254586.1 | 5 - 6 | 777 | 70 | |
XM_011521752.2 | XP_011520054.2 | 5 - 6 | 856 | 70 | |
XM_011521753.2 | XP_011520055.2 | 5 - 6 | 857 | 70 | |
XM_011521754.2 | XP_011520056.1 | 5 - 6 | 487 | 70 | |
XM_017022394.1 | XP_016877883.1 | 5 - 6 | 609 | 70 | |
XM_017022395.1 | XP_016877884.1 | 5 - 6 | 387 | 70 | |
GenBank mRNA | AB046781.2 | - | 5 - 6 | 423 | 70 |
AK055435.1 | - | 5 - 6 | 777 | 70 | |
AL834243.1 | - | 5 - 6 | 752 | 70 | |
BC016881.1 | - | 5 - 6 | 739 | 70 | |
BC113407.1 | - | 5 - 6 | 446 | 70 | |
BC113409.1 | - | 5 - 6 | 446 | 70 | |
BC143266.1 | - | 5 - 6 | 446 | 70 | |
BC143267.1 | - | 5 - 6 | 446 | 70 | |
CN303461.1 | - | 5 - 6 | 528 | 70 |