The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053043.2 | NP_444271.2 | 18 - 18 | 7766 | 76 |
XM_005249531.1 | XP_005249588.1 | 19 - 19 | 7934 | 76 | |
XM_005249532.1 | XP_005249589.1 | 19 - 19 | 7931 | 76 | |
XM_005249533.1 | XP_005249590.1 | 19 - 19 | 7904 | 76 | |
XM_005249534.1 | XP_005249591.1 | 18 - 18 | 7814 | 76 | |
XM_005249535.1 | XP_005249592.1 | 18 - 18 | 7784 | 76 | |
XM_011515840.1 | XP_011514142.1 | 18 - 18 | 7724 | 76 | |
GenBank mRNA | AK055460.1 | - | 1 - 1 | 150 | 76 |
AL832196.1 | - | 8 - 8 | 4499 | 76 |