Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTURN
Entrez Gene ID
222166
Gene Name
maturin, neural progenitor differentiation regulator homolog (Xenopus)
Gene Aliases
C7orf41, Ells1
Chromosome Location
Chr.7: 30134936 - 30162765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152793.2 NP_690006.2 3 - 3 2858 112
XM_005249652.3 XP_005249709.1 4 - 4 1411 112
GenBank mRNA AK055466.1 - 1 - 1 252 112
AK294982.1 - 4 - 4 1198 112
AL834391.1 - 3 - 3 2795 112
AY054121.1 - 3 - 3 2654 112

View Full Product Details