Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB34
Entrez Gene ID
403341
Gene Name
zinc finger and BTB domain containing 34
Gene Aliases
ZNF918
Chromosome Location
Chr.9: 126860665 - 126885878 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099270.1 NP_001092740.1 2 - 2 4458 68
XM_005251989.4 XP_005252046.1 3 - 3 4571 68
XM_011518699.2 XP_011517001.1 2 - 2 4474 68
GenBank mRNA AB082524.1 - 2 - 2 4458 68
AK055661.1 - 1 - 1 252 68
AK074274.1 - 1 - 1 109 68

View Full Product Details