The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205317.1 | NP_001192246.1 | 6 - 6 | 2621 | 88 |
NM_182970.3 | NP_892015.1 | 6 - 6 | 2618 | 88 | |
GenBank mRNA | AK055698.1 | - | 1 - 1 | 151 | 88 |
AK124626.1 | - | 2 - 2 | 2012 | 88 | |
AK126807.1 | - | 6 - 6 | 2521 | 88 | |
BX648947.1 | - | 5 - 5 | 2388 | 88 |