The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000486.5 | NP_000477.1 | 4 - 4 | 2077 | 87 |
GenBank mRNA | AK055824.1 | - | 1 - 1 | 152 | 87 |
BC042496.1 | - | 4 - 4 | 2072 | 87 |