The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145794.1 | NP_001139266.1 | 2 - 3 | 987 | 106 |
NM_001286780.1 | NP_001273709.1 | 2 - 3 | 150 | 106 | |
NM_001286781.1 | NP_001273710.1 | 2 - 3 | 687 | 106 | |
NM_058172.5 | NP_477520.2 | 2 - 3 | 987 | 106 | |
XM_011531587.2 | XP_011529889.1 | 2 - 3 | 150 | 106 | |
XM_017007711.1 | XP_016863200.1 | 2 - 3 | 687 | 106 | |
GenBank mRNA | AB209913.1 | - | 2 - 3 | 575 | 106 |
AY040326.1 | - | 2 - 3 | 364 | 106 | |
AY233452.1 | - | 2 - 3 | 227 | 106 | |
BC107876.1 | - | 2 - 3 | 153 | 106 | |
BC114374.1 | - | 2 - 3 | 99 | 106 |