Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COG7
Entrez Gene ID
91949
Gene Name
component of oligomeric golgi complex 7
Gene Aliases
CDG2E
Chromosome Location
Chr.16: 23388493 - 23453215 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153603.3 NP_705831.1 13 - 14 2001 69
XM_017023870.1 XP_016879359.1 13 - 14 2044 69
GenBank mRNA AF070568.1 - 5 - 6 573 69
AY358632.1 - 8 - 9 1038 69
BC000549.2 - 13 - 14 1956 69
BC037563.1 - 13 - 14 1990 69
KU178837.1 - 13 - 14 1804 69

View Full Product Details