The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037806.3 | NP_001032895.2 | 8 - 9 | 3300 | 84 |
XM_005269050.3 | XP_005269107.1 | 8 - 9 | 3625 | 84 | |
XM_006719525.2 | XP_006719588.1 | 8 - 9 | 3595 | 84 | |
XM_011538610.2 | XP_011536912.1 | 9 - 10 | 3379 | 84 | |
XM_011538611.2 | XP_011536913.1 | 9 - 10 | 3349 | 84 | |
GenBank mRNA | AB046822.1 | - | 1 - 2 | 2241 | 84 |
AF370366.1 | - | 1 - 2 | 84 | ||
BC007998.1 | - | 2 - 3 | 819 | 84 | |
BC033253.1 | - | 1 - 2 | 1816 | 84 |