Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLMP
Entrez Gene ID
79827
Gene Name
CXADR like membrane protein
Gene Aliases
ACAM, ASAM, CSBM, CSBS
Chromosome Location
Chr.11: 123072006 - 123195305 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024769.3 NP_079045.1 1 - 2 391 60
XM_011542998.2 XP_011541300.1 1 - 2 359 60
GenBank mRNA AK026068.1 - 1 - 2 367 60
AK304843.1 - 1 - 2 338 60
AY326422.1 - 1 - 2 215 60
AY358340.1 - 1 - 2 215 60
BC009371.2 - 1 - 2 37 60
BE272517.1 - 1 - 2 284 60
CA395814.1 - 1 - 2 304 60

View Full Product Details