The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_171998.3 | NP_741995.1 | 1 - 2 | 519 | 95 |
GenBank mRNA | AL834460.1 | - | 1 - 2 | 382 | 95 |
BC009714.1 | - | 1 - 2 | 519 | 95 |