Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DNAJC5
- Entrez Gene ID
-
80331
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C5
- Gene Aliases
-
CLN4, CLN4B, CSP, DNAJC5A, NCL, mir-941-2, mir-941-3, mir-941-4, mir-941-5
- Chromosome Location
-
Chr.20: 63895102 - 63936031 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK128776.1 |
- |
5 - 6 |
1460 |
69 |
View Full Product Details