Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZSCAN20
Entrez Gene ID
7579
Gene Name
zinc finger and SCAN domain containing 20
Gene Aliases
KOX29, ZFP-31, ZNF31, ZNF360
Chromosome Location
Chr.1: 33472591 - 33504253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006710876.3 XP_006710939.1 3 - 4 758 61
GenBank mRNA BC011404.2 - 2 - 3 738 61

View Full Product Details