The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015261.2 | NP_056076.1 | 24 - 25 | 3709 | 56 |
XM_011542719.2 | XP_011541021.1 | 23 - 24 | 2970 | 56 | |
GenBank mRNA | AK025667.1 | - | 8 - 9 | 1000 | 56 |
AK124878.1 | - | 24 - 25 | 3709 | 56 | |
BC011408.1 | - | 2 - 3 | 284 | 56 | |
BC098398.1 | - | 24 - 25 | 3168 | 56 | |
BC144524.1 | - | 25 - 26 | 3534 | 56 | |
D29954.1 | - | 24 - 25 | 3131 | 56 |