The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138492.5 | NP_612501.3 | 1 - 2 | 191 | 63 |
GenBank mRNA | BC012033.1 | - | 1 - 2 | 158 | 63 |