The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003196.2 | NP_003187.1 | 4 - 5 | 531 | 84 |
XM_006710864.1 | XP_006710927.1 | 4 - 5 | 504 | 84 | |
XM_011542053.2 | XP_011540355.1 | 4 - 5 | 504 | 84 | |
XM_017002200.1 | XP_016857689.1 | 4 - 5 | 504 | 84 | |
XM_017002201.1 | XP_016857690.1 | 3 - 4 | 465 | 84 | |
XM_017002203.1 | XP_016857692.1 | 4 - 5 | 591 | 84 | |
GenBank mRNA | AJ223473.1 | - | 3 - 4 | 301 | 84 |
AK027024.1 | - | 4 - 5 | 492 | 84 | |
AK290272.1 | - | 4 - 5 | 528 | 84 | |
AK310039.1 | - | 3 - 4 | 465 | 84 | |
AY540752.1 | - | 4 - 5 | 459 | 84 | |
BC041613.1 | - | 4 - 5 | 414 | 84 | |
BQ949868.1 | - | 4 - 5 | 543 | 84 |