Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169513636-169514155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F5
Entrez Gene ID
2153
Gene Name
coagulation factor V
Gene Aliases
FVL, PCCF, RPRGL1, THPH2
UniGene
Hs.30054
Gene Chromosome Location
Chr.1: 169511954-169586632 on Build GRCh38
Transcripts
NM_000130.4 XM_017000660.1

SNP Detail

SNP ID Chromosome Location
rs1041256521 Chr.1: 169513782-169513782 on Build GRCh38
rs537106464 Chr.1: 169513955-169513955 on Build GRCh38
rs866993409 Chr.1: 169513895-169513895 on Build GRCh38
rs780041368 Chr.1: 169513753-169513753 on Build GRCh38
rs1000726290 Chr.1: 169513910-169513910 on Build GRCh38
rs76510731 Chr.1: 169513937-169513937 on Build GRCh38
rs116921763 Chr.1: 169514025-169514025 on Build GRCh38
rs886045537 Chr.1: 169513913-169513913 on Build GRCh38
rs970343110 Chr.1: 169513920-169513920 on Build GRCh38
rs926258080 Chr.1: 169513716-169513716 on Build GRCh38
rs1040892944 Chr.1: 169514044-169514044 on Build GRCh38
rs901104737 Chr.1: 169513787-169513787 on Build GRCh38
rs896154589 Chr.1: 169513853-169513853 on Build GRCh38
rs778934920 Chr.1: 169514008-169514008 on Build GRCh38
rs72708017 Chr.1: 169513837-169513837 on Build GRCh38
rs544753372 Chr.1: 169513715-169513715 on Build GRCh38
rs370721761 Chr.1: 169514111-169514111 on Build GRCh38
rs921971116 Chr.1: 169513814-169513814 on Build GRCh38
rs776516137 Chr.1: 169513799-169513799 on Build GRCh38
rs867675305 Chr.1: 169513989-169513989 on Build GRCh38
rs940984490 Chr.1: 169514031-169514031 on Build GRCh38
rs115882472 Chr.1: 169513950-169513950 on Build GRCh38
rs948962437 Chr.1: 169513701-169513701 on Build GRCh38
rs908308215 Chr.1: 169513976-169513976 on Build GRCh38
rs917533119 Chr.1: 169513665-169513665 on Build GRCh38
rs900964932 Chr.1: 169514049-169514049 on Build GRCh38
rs966076284 Chr.1: 169513959-169513959 on Build GRCh38
rs886045538 Chr.1: 169513923-169513923 on Build GRCh38
rs756353736 Chr.1: 169513833-169513833 on Build GRCh38
rs180742904 Chr.1: 169513732-169513732 on Build GRCh38
rs771034271 Chr.1: 169513946-169513946 on Build GRCh38
rs200174120 Chr.1: 169513662-169513662 on Build GRCh38
rs746426743 Chr.1: 169514013-169514013 on Build GRCh38
rs188490117 Chr.1: 169513872-169513872 on Build GRCh38
rs186962725 Chr.1: 169514097-169514097 on Build GRCh38
rs1030939168 Chr.1: 169513685-169513685 on Build GRCh38
rs368526761 Chr.1: 169514030-169514030 on Build GRCh38
rs1049710061 Chr.1: 169514118-169514118 on Build GRCh38
rs548119385 Chr.1: 169513864-169513864 on Build GRCh38
rs993031440 Chr.1: 169513897-169513897 on Build GRCh38
rs531400760 Chr.1: 169513743-169513743 on Build GRCh38
rs746804474 Chr.1: 169513800-169513800 on Build GRCh38
rs908042169 Chr.1: 169514027-169514027 on Build GRCh38
rs558628625 Chr.1: 169513992-169513992 on Build GRCh38
rs920842983 Chr.1: 169513929-169513929 on Build GRCh38
rs183726377 Chr.1: 169513759-169513759 on Build GRCh38
rs1049666332 Chr.1: 169513827-169513827 on Build GRCh38
rs75302539 Chr.1: 169513832-169513832 on Build GRCh38
rs903569717 Chr.1: 169513909-169513909 on Build GRCh38
rs181328696 Chr.1: 169513917-169513917 on Build GRCh38
rs930835327 Chr.1: 169513819-169513819 on Build GRCh38
rs956639806 Chr.1: 169513923-169513923 on Build GRCh38
rs536423692 Chr.1: 169513927-169513927 on Build GRCh38
rs542871939 Chr.1: 169513721-169513721 on Build GRCh38
rs1044830658 Chr.1: 169513887-169513887 on Build GRCh38

View Full Product Details